Skip to Content
Merck
All Photos(1)

Key Documents

CP26

Sigma-Aldrich

Anti-Integrin β₁ Mouse mAb (4B7)

liquid, clone 4B7, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody product type

primary antibodies

clone

4B7, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

should not react with

mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

General description

Purified mouse monoclonal antibody generated by immunizing mice with the specified immunogen and fusing splenocytes with Sp2/0 mouse myeloma cells. Recognizes the β1 integrin subunit.
Recognizes the β1 integrin subunit in A431 and Jurkat cells.

Immunogen

Human
recombinant, human β1 integrin

Application

Frozen Sections (2 µg/ml)

Immunofluorescence (2.5 µg/ml)

Immunoprecipitation (1 µg/sample)

Paraffin Sections (2 µg/ml, no pre-treatment required)

Immunoblotting (not recommended)

Neutralization Studies (see application references)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM sodium phosphate buffer, 50% glycerol.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
A-431 or Jurkat cells or skin tissue

Other Notes

Akiyama, S.K. 1996. Hum. Cell.9, 181.
Farnoud, M.R. et al. 1996. Int. J. Cancer67, 45.
Morimoto, C. and Tachibana, K. 1996. Hum. Cell.9, 163.
Webb, D.L. et al. 1996. J. Cell. Biochem.61, 127.
Schaller, M.D. et al. 1995. J. Cell. Biol.130, 1181.
Akiyama, S.K. et al. 1994. J. Biol. Chem.269, 15961.
Albelda, S.M. 1993. Lab. Invest.68, 4.
Evans, C.W. 1992. Cell. Biol. Int. Rep.16, 1.
Albelds, S.M. and Buck, C.A. 1990. FASEB J.4, 2868.
Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang-Er Chen et al.
Scientific reports, 7(1), 12718-12718 (2017-10-07)
Reversible phosphorylation of proteins and the assembly of thylakoid complexes are the important protective mechanism against environmental stresses in plants. This research was aimed to investigate the different responses of the antioxidant defense system and photosystem II (PSII) to osmotic
Anna L Guildford et al.
Journal of the Royal Society, Interface, 8(58), 641-649 (2010-11-26)
In-stent restenosis is a clinical complication following coronary angioplasty caused by the implantation of the metal device in the atherosclerotic vessel. Histological examination has shown a clear contribution of both inflammatory and smooth muscle cells (SMCs) to the deposition of
Ana Carreras-González et al.
PLoS pathogens, 15(11), e1008163-e1008163 (2019-11-19)
Macrophages mediate the elimination of pathogens by phagocytosis resulting in the activation of specific signaling pathways that lead to the production of cytokines, chemokines and other factors. Borrelia burgdorferi, the causative agent of Lyme disease, causes a wide variety of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service