CN03F2001
Polygard® CN Cartridge Filter
pore size 0.3 μm, Code F (double open end w/flat gaskets)
Synonym(s):
CN CARTRIGE C-F 20" 0.3UM
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
polypropylene
pore size
0.3 μm nominal pore size
0.3 μm pore size
cartridge code
Code F (double open end w/flat gaskets)
Looking for similar products? Visit Product Comparison Guide
General description
Device Configuration: Cartridge
Legal Information
POLYGARD is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service