Skip to Content
Merck
All Photos(1)

Key Documents

AP505F

Sigma-Aldrich

Goat Anti-Human Ig κ chain Antibody, FITC conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€280.00

€280.00


Estimated to ship on02 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€280.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€280.00


Estimated to ship on02 May 2025


Request a Bulk Order

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

General description

Antibody molecules typically comprise two immunoglobulin light chains covalently bound to a pair of heavy chains. Immunoglobulin light chains occur in two types, designated by the Greek letters kappa and lambda. Kappa and lambda light chains are approximately 250 amino acids in length with an average mass of about 25 kDa. The ratio of kappa to lambda found in the immunoglobulin population varies by species.

Specificity

Reacts with human kappa light chains. Absorbed for human myeloma proteins with λ light chains.

Application

Immunofluorescence: ≤1 μg/106 cells.

Optimal working dilutions must be determined by the end user.
This Goat anti-Human Ig κ chain Antibody, FITC conjugate is validated for use in IF for the detection of Human Ig κ chain.

Physical form

Purified by affinity chromatography on pooled human Igs with kappa light chains covalently linked to agarose. Liquid in 1.0 mL PBS/NaN3.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joel G Turner et al.
Oncotarget, 7(48), 78896-78909 (2016-11-03)
Acquired proteasome-inhibitor (PI) resistance is a major obstacle in the treatment of multiple myeloma (MM). We investigated whether the clinical XPO1-inhibitor selinexor, when combined with bortezomib or carfilzomib, could overcome acquired resistance in MM. PI-resistant myeloma cell lines both in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service