Skip to Content
Merck
All Photos(1)

Key Documents

AP501A

Sigma-Aldrich

Goat Anti-Mouse IgM+IgG+IgA (H+L) Antibody, Alkaline Phosphatase, Species Adsorbed

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€321.00

€321.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
€321.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

€321.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

shipped in

dry ice

target post-translational modification

unmodified

General description

IgG is the most abundant immunoglobulin compose of two heavy chains and two light chains. Each molecule has two antigen binding sites. IgG has 4 subclasses: IgG1 (66%), IgG2 (23%), IgG3 (7%) and IgG4 (4%). IgM constitutes about 10% of serum immunoglobulins. IgM (with IgD) is the major immunoglobulin expressed on the surface of B cells. Monomeric IgA constitutes 5-15 % of the serum immunoglobulins whereas dimeric IgA is localized to mucosa surfaces.

Specificity

Reacts with heavy and light chains of mouse IgM, IgG1, IgG2a, IgG2b, IgG3 and IgA. Absorbed for human sera and purified human paraproteins. Minimal cross reactivity with human immunoglobulins.

Application

ELISA: 1:2,000-1:4,000

Optimal working dilutions must be determined by the end user.
Goat anti-Mouse IgM+IgG+IgA (H+L) Antibody, Alkaline Phosphatase, Species Adsorbed detects level of Mouse IgM+IgG+IgA & has been published & validated for use in ELISA.

Physical form

Purified by affinity chromatography on pooled mouse IgM+IgG+IgA covalently linked to agarose. Liquid in 1.0 mL of stock solution in 50mM Tris/1mM MgCl2/50% Glycerol, pH 8.0, containing 0.1% NaN3 as preservative.

Storage and Stability

Maintain refrigerated at 2°-8°C under sterile conditions for up to twelve months from date of receipt. Store long term at -20°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stuart D Dowall et al.
PLoS neglected tropical diseases, 11(7), e0005704-e0005704 (2017-07-04)
Zika virus (ZIKV) falls into two lineages: African (ZIKVAF) and Asian (ZIKVAS). These lineages have not been tested comprehensively in parallel for disease progression using an animal model system. Here, using the established type-I interferon receptor knockout (A129) mouse model

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service