Skip to Content
Merck
All Photos(1)

Key Documents

AP108C

Sigma-Aldrich

Goat Anti-Guinea Pig IgG Antibody, Cy3 conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
€309.00

€309.00


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
2 MG
€309.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

€309.00


Estimated to ship on12 April 2025


Request a Bulk Order

biological source

goat

Quality Level

conjugate

CY3 conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

guinea pig

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Application

Goat anti-Guinea Pig IgG Antibody, Cy3 conjugate detects level of Guinea Pig IgG & has been published & validated for use in IF.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Physical form

Lyophilized from 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, 15 mg/mL Bovine Serum Albumin (IgG-Free, Protease-Free), and 0.05% Sodium Azide.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony Avellino et al.
International journal of molecular sciences, 24(15) (2023-08-12)
Cell cycle progression during development is meticulously coordinated with differentiation. This is particularly evident in the Drosophila 3rd instar eye imaginal disc, where the cell cycle is synchronized and arrests at the G1 phase in the non-proliferative region (NPR), setting
X chromosome control of meiotic chromosome synapsis in mouse inter-subspecific hybrids.
Bhattacharyya, T; Reifova, R; Gregorova, S; Simecek, P; Gergelits, V; Mistrik et al.
PLoS Genetics null
David Homolka et al.
Chromosoma, 121(1), 91-104 (2011-10-18)
During meiosis, pairing of homologous chromosomes and their synapsis are essential prerequisites for normal male gametogenesis. Even limited autosomal asynapsis often leads to spermatogenic impairment, the mechanism of which is not fully understood. The present study was aimed at deliberately
Andrew S Lapato et al.
ASN neuro, 12, 1759091420979604-1759091420979604 (2020-12-11)
While seizure disorders are more prevalent among multiple sclerosis (MS) patients than the population overall and prognosticate earlier death & disability, their etiology remains unclear. Translational data indicate perturbed expression of astrocytic molecules contributing to homeostatic neuronal excitability, including water
Natalie R Powers et al.
Science advances, 6(43) (2020-10-25)
In many mammals, genomic sites for recombination are determined by the histone methyltransferase PRMD9. Some mouse strains lacking PRDM9 are infertile, but instances of fertility or semifertility in the absence of PRDM9 have been reported in mice, canines, and a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service