Skip to Content
Merck
All Photos(1)

Key Documents

ABT1372

Sigma-Aldrich

Anti-phospho-Keratin-8 Antibody (Ser23)

from rabbit, purified by affinity chromatography

Synonym(s):

Keratin, type II cytoskeletal 8, Ser23-phosphorylated, CK-8, Ser23-phosphorylated, Cytokeratin-8, Ser23-phosphorylated, K8, Ser23-phosphorylated, Keratin-8, Ser23-phosphorylated, Type-II keratin Kb8, Ser23-phosphorylated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology)

technique(s)

inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer23)

Gene Information

human ... KRT8(3856)

General description

Keratin, type II cytoskeletal 8 (UniProt P05787; also known as CK-8, Cytokeratin-8, K8, Keratin-8, Type-II keratin Kb8) is encoded by the KRT8 (also known as CYK8) gene (Gene ID 3856) in human. Cytokeratin-8 (Keratin-8 or K8) belongs to a family of highly conserved epithelial-specific intermediate filament (IF) proteins that play an important role in the mechanical stability and integrity of epithelial cells and tissues. Each epithelium is characterized by its unique keratin expression pattern. Keratin 8 is a type II intermediate filament protein found in simple epithelia, where it pairs with keratin 18 (K8/KI8) and plays an important role in protecting cells from stress, injury, and apoptosis. The structure and function of K8/18 are regulated by posttranslational modifications, including glycosylation, ubiquitination, and phosphorylation. K8 has been reported to be phosphorylated on Ser8, Ser23, Ser73, and Ser431 by various protein kinases. PKCepsilon is reported to mediate K8 phosphorylation on Ser8 and Ser23, p38 and PKCdelta are responsible for K8 Ser73 phosphorylation, while ERK and JNK are involved in K8 Ser431 phosphorylation.

Specificity

This rabbit polyclonal antibody detected the target phospho band only in lysates from phosphatase inhibitors-treated, but not untreated HeLa cells. Antibody blocking by the immunogen peptide, but not the corresponding nonphosphorylated peptide prior to immunoblotting abolished target band detection. The Ser23 designation is based on human keratin-8 sequence with initiator Met1 removed. The phosphorylation site sequence in mouse/rat (-SSR(pS)YT-) differs from that of human (-SSR(pS)FT-) by a single Phe24-to-Tyr24 switch (F24Y). Cross-reactivity of this polyclonal antibody toward murine and rat species is likely, but has not been confirmed.

Immunogen

KLH-conjugated linear peptide corresponding to the phosphorylation site sequence containing the phosphorylated target serine.

Application

Peptide Inhibition Assay: Preincubation of a representative lot with the immunogen phosphopeptide, but not the corresponding nonphosphopeptide, abolished target band detection by Western blotting analysis of 10 µg lysate from phosphatase inhibitors-treated HeLa cells.
Research Category
Cell Structure
This rabbit polyclonal Anti-phospho-Keratin-8 (Ser23), Cat. No. ABT1372 is validated for use in Peptide Inhibition Assay and Western Blotting for the detection of Keratin 8 phosphorylated at serine 23.

Quality

Evaluated by Western Blotting in HeLa cells lysate.

Western Blotting Analysis: A 1:1,000 dilutiion of this antibody detected keratin-8 Ser23 phosphorylation in 10 µg of lysate from phosphatase inhibitors-treated, but not untreated, HeLa cells.

Target description

~54 kDa observed. 53.57/56.48 kDa (isoform 1/2) calculated (Met1 removed). Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service