Skip to Content
Merck
All Photos(2)

Key Documents

ABS2252

Sigma-Aldrich

Anti-ZnT8

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€151.00
100 μL
€381.00

€151.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
€151.00
100 μL
€381.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€151.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

mol wt

calculated mol wt 40.76 kDa

species reactivity

mouse, human

packaging

antibody small pack of 100 μL

technique(s)

immunofluorescence: suitable

isotype

IgG

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

General description

Zinc transporter 8 (UniProt: Q8IWU4; also known as ZnT-8, Solute carrier family 30 member 8) is encoded by the SLC30A8 (also known as ZNT8) gene (Gene ID: 169026) in human. Zinc is required for insulin crystallization, storage, and secretion in the secretory granules of pancreatic β-cells. Delivery of Zn2+ into the insulin-containing granules is largely accomplished by the ZnT8 protein. ZnT8 is a homodimeric, multi-pass membrane protein with four cytoplasmic domains, six transmembrane domains, and three extracellular domains. It serves as a granule-specific Zn2+ transporter that is abundantly expressed in pancreatic islet cells. It contains a histidine-rich region, HXXXXXHNH-motif, in its third cytoplasmic domain, which serves as a ligand for zinc. ZnT8 is expressed in both a and b cells, but not in  cells. It plays a major role in Zn2+ uptake into the secretory granules. During stimulated secretion, Zn2+ is shown to be co-released with other granular content into the extracellular medium.Loss of ZnT8 function is reported to reduce total islet Zn2+ content as well as free Zn2+ in the cytosol and granules. Loss of ZnT8 also results in reduced dense core granules and increased granule diameter. Studies have shown that the expression of ZnT8 is significantly reduced in the islets of diabetic murine models. Two isoforms of ZnT8 have been described that are produced by alternative splicing. (Ref.: Ghazvini Zadeh, EH., et al. (2020). Cell Reports 32(2); 107904; Dwivedi, OP., et al. (2019). Nat. Genetics. 51(11); 1596-1606).

Specificity

This rabbit polyclonal antibody detects Zinc transporter 8 (ZnT8). It targets an epitope within the C-terminal, cytoplasmic domain.

Immunogen

GST-tagged recombinant fragment corresponding to 102 amino acids from the C-terminal, cytoplasmic domain of human zinc transporter 9 (ZnT8).

Application

Quality Control Testing

Evaluated by Immunofluorescence in mouse pancreas frozen formalin fixed sections.

Immunofluorescence Analysis: A 1:100 dilution of this antibody detected ZnT8 in mouse pancreas frozen formalin fixed sections.

Tested Applications


Immunofluorescence Analysis: A representative lot detected ZnT8 in Immunofluorescence applications (Zadeh, EHG., et.al. (2020). Cell Rep. 32(2):107904).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-ZnT8, Cat. No. ABS2252, is a rabbit polyclonal antibody that detects Zinc transporter 8 (ZnT8) and is tested in Immunofluorescence.

Physical form

Rabbit polyclonal antibody in 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service