Skip to Content
Merck
All Photos(1)

Key Documents

ABS192

Sigma-Aldrich

Anti-phospho-SRPK2 (Ser497) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

SRSF protein kinase 2, SFRS protein kinase 2, Serine/arginine-rich protein-specific kinase 2, SR-protein-specific kinase 2, SRSF protein kinase 2 N-terminal, SRSF protein kinase 2 C-terminal

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€374.00

€374.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
€374.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€374.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

hamster, mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), Xenopus (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer497)

Gene Information

human ... SRPK2(6733)

General description

SRPK2 is also known as SRSF protein kinase 2, SFRS protein kinase 2, and Serine/arginine-rich protein-specific kinase 2. SRPK2 is a kinase which is involved in the phosphorylation of SR splicing factors, and the regulation of splicing. SRPK2 also promotes neuronal apoptosis, plays an essential role in splicesomal B complex formation, and mediates hepatitis B virus (HBV) replication and core protein phosphorylation. SRPK2 is highly expressed in brain, moderately expressed in heart and skeletal muscle and at low levels in lung, liver, and kidney.

Specificity

This antibody recognizes the protein kinase domain of SRPK2 phosphorylated at Ser497.

Immunogen

Epitope: Protein kinase domain
KLH-conjugated linear peptide corresponding to the protein kinase domain of human SRPK2 (Ser497).

Application

Anti-phospho-SRPK2 (Ser497) Antibody is a highly specific rabbit polyclonal antibody, that targets SRPK & has been tested in western blotting.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in NIH3T3 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected SRPK2 (Ser497) in 10 µg of NIH3T3 cell lysate and Insulin treated NIH3T3 cell lysate.

Target description

~120 kDa observed. Uniprot describes 2 isoforms produced by alternative splicing at ~77 kDa and ~79 kDa Uniprot states 8 phosphorylation sites are known to exist.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
NIH3T3 cell lysate and Insulin treated NIH3T3 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taylor Ludman et al.
Molecular pain, 15, 1744806919837429-1744806919837429 (2019-02-28)
Chemotherapy-induced painful peripheral neuropathy (CIPN) is the most common toxicity associated with widely used chemotherapeutics. CIPN is the major cause of dose reduction or discontinuation of otherwise life-saving treatment. Unfortunately, CIPN can persist in cancer survivors, which adversely affects their

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service