Skip to Content
Merck
All Photos(2)

Key Documents

ABN98

Sigma-Aldrich

Anti-Neurexin-1-alpha Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Neurexin-1-alpha, Neurexin I-alpha, Neurexin1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€500.00

€500.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€500.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€500.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), canine (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NRXN1(9378)

General description

Neurexins are heavily glycosylated and processed neuronal transmembrane cell surface proteins likely involved in cell recognition and adhesion (see review by Missler & Sudhof, 1998). Antibody Anti-Neurexin-1-alpha (ABN35) was made against a peptide immunogen corresponding to a stretch of amino acids in the extracellular domain of human Neurexin-1-alpha (Uniprot Q9ULB1). Although the theoretical full length of Neurexin-1-alpha is 161 kDa, the protein detected in human brain lysates may show a band around 160 kDa and additional bands around 45 kDa as a possible result of the posttranslational processing previously reported.

Specificity

This antibody recognizes Neurexin-1-alpha at the extracellular laminin G-like 3 domain.

Immunogen

Epitope: Extracellular laminin G-like 3 domain
KLH-conjugated Linear peptide corresponding to human Neurexin-1-alpha at the extracellular laminin G-like 3 domain.

Application

Detect Neurexin-1-alpha using this Anti-Neurexin-1-alpha Antibody validated for use in WB, IH(P).
Immunohistochemistry Analysis: A 1:100 dilution from a representative lot detected Neurexin-1-alpha in human brain tissue.
Research Category
Neuroscience
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Western Blot in human cerebellum tissue lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected Neurexin-1-alpha on 10 µg of human cerebellum tissue lysate.

Target description

~ 45 kDa Observed At higher concentrations or longer exposures, in certain lysates, a band at ~150kD can be seen that corresponds to the expected full length Mw.
UniProtKB/Swiss-Prot Q9ULB1 (NRX1A_HUMAN) entry describes 3 isoforms produced by alternative promoter usage and alternative splicing. Reference (Accolade/Related Documents) TIG JANUARY 1998 VOL. 14 No. 1 indicates that multiple neurexin isoforms are generated from three genes by alternative promoters and alternative splicing.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human cerebellum tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service