Skip to Content
Merck
All Photos(3)

Key Documents

ABN27

Sigma-Aldrich

Anti-GRIP-1 Antibody, CT

from rabbit, purified by affinity chromatography

Synonym(s):

glutamate receptor interacting protein 1, glutamate receptor-interacting protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€383.00

€383.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€383.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€383.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), human (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Glutamate receptor interacting protein-1 (GRIP-1) is a multi-PDZ domain scaffold protein and a member of the steroid receptor coactivator (SRC) family of proteins. GRIP-1 interacts and maneuvers kinesin heavy chains to dendrites by acting as the power source for AMPA receptors. During initial stages of embryonic development, GRIP-1 is essential for typical cell-matrix interactions.

Specificity

This antibody recognizes GRIP-1 at the C-terminus.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of rat GRIP-1.

Application

Detect GRIP-1 using this Anti-GRIP-1 Antibody, C-terminus validated for use in WB, IP, IH(P).
Immunoprecipitation Analysis: 10 µg from a previous lot immunoprecipitated GRIP-1 from rat brain cytosol tissue lysate.

Immunohistochemistry Analysis: 1:300 dilution from a previous lot detected GRIP-1 in rat brain cells.
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Western Blot in rat brain cytosol tissue lysate.

Western Blot Analysis: 0.05 µg/mL of this antibody detected GRIP-1 on 10 µg of rat brain cytosol tissue lysate.

Target description

~121 kDa observed

Linkage

Replaces: 06-986

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain cytosol tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisabetta Gerace et al.
Journal of neurochemistry (2020-10-28)
Modifications in the subunit composition of AMPA receptors (AMPARs) have been linked to the transition from physiological to pathological conditions in a number of contexts, including EtOH-induced neurotoxicity. Previous work from our laboratory showed that EtOH withdrawal causes CA1 pyramidal
Rocío Palenzuela et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 37(41), 9945-9963 (2017-09-15)
The regulated transport of AMPA-type glutamate receptors (AMPARs) to the synaptic membrane is a key mechanism to determine the strength of excitatory synaptic transmission in the brain. In this work, we uncovered a new role for the microtubule-associated protein MAP1B
Manjeet K Rao et al.
The Journal of biological chemistry, 289(51), 35087-35101 (2014-10-22)
Genome-wide studies have revealed that genes commonly have a high density of RNA polymerase II just downstream of the transcription start site. This has raised the possibility that genes are commonly regulated by transcriptional elongation, but this remains largely untested

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service