Skip to Content
Merck
All Photos(1)

Documents

ABN255

Sigma-Aldrich

Anti-Bassoon Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

affinity purified immunoglobulin

Quality Level

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rabbit, rat

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

General description

GST-tagged recombinant protein corresponding to mouse Bassoon.

Specificity

This antibody is predicted to cross react with both isoforms.

Application

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Bassoon in rat cerebellum, human cerebral cortex, rat colon, mouse small intestine, and rat kidney tissue.

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected Bassoon in 10 µg of mouse brain tissue lysate.

Physical properties

~420 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service