Skip to Content
Merck
All Photos(5)

Key Documents

ABN1464

Sigma-Aldrich

Anti-GRP-78

from rabbit, purified by affinity chromatography

Synonym(s):

78 kDa glucose-regulated protein, GRP-78, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, Heat shock 70 kDa protein 5, Immunoglobulin heavy chain-binding protein, BiP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Pricing and availability is not currently available.

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

ground squirrel (based on 100% sequence homology), hamster (based on 100% sequence homology), bovine (based on 100% sequence homology), mouse (based on 100% sequence homology), goat (based on 100% sequence homology), primate (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... HSPA5(3309)

General description

78 kDa glucose-regulated protein (UniProt: P11021; also known as GRP-78, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, Heat shock 70 kDa protein 5, Immunoglobulin heavy chain-binding protein, BiP) is encoded by the HSPA5 (also known as GRP78) gene (Gene ID: 3309) in human. GRP78 a member of the HSP family of molecular chaperones that is required for stress-induced autophagy and for endoplasmic reticulum (ER) integrity. GRP78 is involved in multiple processes, including translocation of the newly formed proteins across the ER membrane, folding and assembly of proteins, and targeting misfolded proteins for ER-associated degradation. It plays a critical role in regulating unfolded protein response (UPR). Following ER stress, GRP78 is released from ER transmembrane signal transducers, such as PKR-like ER kinase (PERK), which leads to the activation of UPR signaling pathways. Once GRP78 is dissociated, PERK is allowed to dimerize and is autophosphorylated and activated. Activated PERK phosphorylates elF2alpha that results in the reduction of protein synthesis. (Ref.:Wang, M., et al. (2009). Antioxid Redox Signal. 11(9): 2307 2316).

Specificity

This polyclonal antibody detects GRP-78 protein in multiple mammalian species. It targets an epitope within 17 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal region of human GRP-78.

Application

Anti-GRP-78, Cat. No. ABN1464, is a highly specific rabbit polyclonal antibody that targets 78 kDa glucose-regulated protein and has been tested in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: 0.2 µg/mL from a representative lot detected GRP-78 in 10 µg of A431 cell lysates.

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected GRP-78 in human cerebellum, human pancreas, human small intestine, and human tonsil tissues.

Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected GRP-78 in NIH/3T3 cells.


Quality

Evaluated by Western Blotting in A431 cell lysates.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected GRP-78 in 10 µg of A431 cell lysates.

Target description

~78 kDa observed; 72.33 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service