Skip to Content
Merck
All Photos(2)

Documents

ABE55

Sigma-Aldrich

Anti-Maf-B Antibody

serum, from rabbit

Synonym(s):

v-maf musculoaponeurotic fibrosarcoma oncogene homolog B (avian), MAFB/Kreisler basic region/leucine zipper transcription factor, transcription factor MafB, Kreisler (mouse) maf-related leucine zipper homolog, Kreisler maf-related leucine zipper homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MAFB(9935)

General description

Transcription factor MafB (Maf-B) belongs to the bZIP family as a part of the MAF subfamily and contains a characteristic bZIP domain. Maf-B can function as either a transcriptional activator or repressor. It regulates erythroid differentiation through its interaction with and negative regulation of EST-1 activity. Maf-B is a critical factor in the differentiation of islet beta cells, alpha and beta cells, macrophage, monocytic, and podocyte cells. It is involved in macrophage expression and maturation of F4/80, and regulates renal development and survival of renal tubules. Interaction with SUMO regulates the transcriptional activity of Maf-B along with its function in determining macrophage fate. Studies have shown that, relative to cell context, Maf-B can act as either a tumor or an oncogene suppressor.

Immunogen

KLH-conjugated linear peptide corresponding to human Maf-B.

Application

Anti-Maf-B Antibody is a Rabbit Polyclonal Antibody for detection of Maf-B also known as v-maf musculoaponeurotic fibrosarcoma oncogene homolog B (avian) & has been validated in WB, ICC.
Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected Maf-B in NIH/3T3 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blot in L6 cell lysate.

Western Blot Analysis: 1:1,000 dilution of this antibody detected Maf-B on 10 µg of L6 cell lysate.

Target description

~ 36 kDa observed

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide
Unpurified

Storage and Stability

Store at -20°C
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
L6 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mounia Tannour-Louet et al.
Nature medicine, 20(7), 715-724 (2014-06-02)
Despite the fact that genitourinary defects are among the most common birth defects in newborns, little is known about their etiology. Here we analyzed children born with congenital genitourinary tract masculinization disorders by array-comparative genomic hybridization, which revealed in 1.35%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service