Skip to Content
Merck
All Photos(4)

Key Documents

ABE1321

Sigma-Aldrich

Anti-MBD6

from rabbit, purified by affinity chromatography

Synonym(s):

Methyl-CpG-binding domain protein 6, Methyl-CpG-binding protein MBD6

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€130.00
100 μG
€318.00

€130.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€130.00
100 μG
€318.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€130.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... MBD6(114785)

General description

Methyl-CpG-binding domain protein 6 (UniProt: Q96DN6; also known as Methyl-CpG-binding protein MBD6) is encoded by the MBD6 (also known as KIAA1887) gene (Gene ID: 114785) in human. The methyl-CpG-binding domain (MBD) proteins are primary candidates for the readout of DNA methylation. MBD proteins require a functional MBD domain and methyl-CpGs, although some MBD proteins can also bind unmethylated DNA. MBD proteins are involved in transcriptional repression by histone modification, nucleosome remodeling or chromatin organization. MBD6 is highly expressed in testes. Its MBD domain is localized in the N-terminal region (aa 11-81) and it also contains proline-rich domains. MBD6 binds to heterochromatin, but does not interact with either methylated or unmethylated DNA. MBD6 is reported to interact directly with the human polycomb deubiquitinase complex, PR-DUB, which catalyzes deubiquitination of H2AK119. Mutations in MBD6 that lead to R883W alteration are reported to cause autism. (Ref.: Cukier, HN., et al. (2012). Autism Res. 5(6); 385-397; Du, Q., et al. (2015). Epigenomics 7(6); 1051-1073).

Specificity

This rabbit polyclonal antibody detects Methyl-CpG-binding domain protein 6 in human and murine cells. It targets an epitope within 16 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal half of human Methyl-CpG-binding domain protein 6.

Application

Anti-MBD6, Cat. No. ABE1321, is a highly specific rabbit polyclonal antibody that targets Methyl-CpG-binding domain protein 6 and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected MBD6 in HeLa, A431, HEK293, and NIH3T3 cells.

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected MBD6 in human skin and human stomach tissue sections.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected MBD6 in SH-SY5Y cell lysate.

Target description

~110 kDa observed; 101.20 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service