Skip to Content
Merck
All Photos(2)

Key Documents

ABC74

Sigma-Aldrich

Anti-IGPR-1 Antibody

1 mg/mL, from rabbit

Synonym(s):

Transmembrane and immunoglobulin domain-containing protein 2, Immunoglobulin and proline-rich receptor 1, IGPR-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€370.00

€370.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€370.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€370.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TMIGD2(126259)

General description

IGPR-1 or Immunoglobulin and proline-rich receptor 1, also called Transmembrane and immunoglobulin domain-containing protein 2 and encoded by the human gene TMIGD2/IGPR1 is a widely expressed epithelial and endothelial membrane protein that plays a role in cell-cell interactions, cell migration and angiogenesis. IGPR-1 regulates cellular morphology, homophilic cell aggregation, and cell-cell interaction. IGPR-1 activity also modulates actin stress fiber formation and focal adhesion and reduces cell migration. Silencing of expression of IGPR-1 by small interfering RNA (siRNA) and by ectopic overexpression in endothelial cells showed that IGPR-1 regulates capillary tube formation in vitro, and in B16F melanoma cells engineered to express IGPR-1 displayed extensive angiogenesis in the mouse Matrigel angiogenesis model. Moreover, IGPR-1, through its proline-rich cytoplasmic domain, associates with multiple Src homology 3 (SH3)-containing signaling proteins, including SH3 protein interacting with Nck (SPIN90/WISH), bullous pemphigoid antigen-1, and calcium channel β2.

Immunogen

Epitope: Cytoplasmic domain
Linear peptide corresponding to the cytoplasmic domain of the human mature form of IGPR-1.

Application

Anti-IGPR-1 antibody is an antibody against IGPR-1 for use in western blotting, ICC & IHC.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: 2 µg/mL from a representative lot detected IGPR-1 in 10 µg of HepG2 cell lysate, human placenta tissue lysate, HUVEC cell lysate, and L9 cell lysate.

Western Blotting Analysis: A representative lot from an independent laboratory detected IGPR-1 in IGPR-1 transfected PAE cell lysates (Rahimi, N., et al. (2012). Mol Cell Biol. 23(9):1646-1556.).

Immunocytochemistry Analysis: A representative lot from an independent laboratory detected IGPR-1 in IGPR-1 transfected PAE cells (Rahimi, N., et al. (2012). Mol Cell Biol. 23(9):1646-1556.).

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected IGPR-1 in human colon cancer tissues (Rahimi, N., et al. (2012). Mol Cell Biol. 23(9):1646-1556.).

Quality

Evaluated by Western Blotting in PC-12 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected IGPR-1 in 10 µg of PC-12 cell lysate.

Target description

~58 kDa observed. Uniprot describes two isoforms at 31 kDa and 30 kDa The pre-mature and mature form of this protein may be observed at 32 kDa and 58 kDa (Rahimi, N., et al. (2012). Mol Cell Biol. 23(9):1646-1556.). Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service