Skip to Content
Merck
All Photos(1)

Key Documents

AB9856

Sigma-Aldrich

Anti-TorsinB Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€435.00

€435.00


Estimated to ship on10 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€435.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€435.00


Estimated to ship on10 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TOR1B(27348)

Specificity

TorsinB. The antibody reacts with the ~38 kDa protein.

Immunogen

Synthetic peptide from torsinB.

Application

Detect TorsinB using this Anti-TorsinB Antibody validated for use in IP, WB & IC.
Western blot: 1:100-1:200. The antibody reacts with the ~38 kDa protein. The suggested blocking buffer is TBS with 10% milk and 0.05% Tween-20. The suggested dilution buffer is TBS with 1% milk and 0.05% Tween-20. Note: TorsinB is a glycoprotein which can be deglycosylated by either EndoH or PNGase treatment.
Immunocytochemistry: 1:100 using rhodamine conjugated goat anti-rabbit secondary antibody. Suggested fixative is 4% paraformaldehyde fixed (37°C for 20 minutes) cells. Suggested blocking solution should contain 10% goat serum. Suggested dilution buffer is PBS containing 1% BSA.
Immunoprecipitation. Recommended antibody amount is 5-7 mL of rabbit serum. The preferred tissue/cell lysis buffer is RIPA. Final reaction volume of 1000 mL. Final total protein concentration in reaction mix: 0.1-0.5 mg/mL. Suggested incubation time is 16-18 hours at 2-8°C. Sugggested capture is Protein A/G.

Optimal working dilutions must be determined by end user.

Storage and Stability

Maintain at frozen at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Western blot: brain and liver. Immunocytochemistry: SH-SY5Y cells.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service