Skip to Content
Merck
All Photos(1)

Key Documents

AB9842

Sigma-Aldrich

Anti-phospho-Munc-18 (Ser515) Antibody

Chemicon®, from rabbit

Synonym(s):

ANC18HA, Munc18-1, n-Sec1, nSsec1, NSEC1A, p67, Protein unc-18 homolog 1, Protein unc-18 homolog A, rbSec1, rbSec1A, rbSec1B, Sec1, Syntaxin-binding protein 1, Unc18-1, Unc-18A, Unc-18 homolog, Stxbp1, Unc18a

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€344.00

€344.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€344.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€344.00


Estimated to ship on01 June 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer515)

Gene Information

Specificity

Munc-18, phospho Serine 515. The antibody recognizes a protein of ~65 kDa corresponding to Munc-18, phospho Serine 515 in lysates from rat cortex. Immunolabeling is blocked by the phosphopeptide used as the immunogen but not the corresponding non-phosphopeptide.
The immunogen has 100% homology with human, mouse, non-human primates, bovine, canine, chicken and Xenopus.

Immunogen

Synthetic peptide corresponds to sequence surrounding the phosphoserine 515 site of rat Munc-18.

Application

This Anti-phospho-Munc-18 (Ser515) Antibody is validated for use in WB for the detection of phospho-Munc-18 (Ser515).

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shatrunjai Giri et al.
Journal of neuroscience research, 99(7), 1815-1834 (2021-04-06)
Rapid eye movement sleep (REMS) favors brain development and memory, while it is decreased in neurodegenerative diseases. REMS deprivation (REMSD) affects several physiological processes including memory consolidation; however, its detailed mechanism(s) of action was unknown. REMS reduces, while REMSD elevates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service