Skip to Content
Merck
All Photos(1)

Key Documents

AB5652P

Sigma-Aldrich

Anti-Histamine Receptor 1 Antibody, CT

Chemicon®, from rabbit

Synonym(s):

H1R

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€412.00

€412.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€412.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€412.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

bovine, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Hrh1(15465)
rat ... Hrh1(24448)

Specificity

Recognizes rat Histamine Receptor 1 (HR1). The immunogen shows no significant sequence homology with other histamine receptors.

The immunogen peptide is 100% conserved in mouse and bovine, 94% in human, fish and monkey and 80% in guinea pig. Reactivity with other species has not been confirmed.

Immunogen

A 17 amino acid peptide sequence near the cytoplasmic, C-terminus of rat HR1 (Fujimoto et al. 1993; Inoue et al. 1996; DeBacker et al. 1993; Fukui et al. 1994; Moguilevsky et al. 1994).
Epitope: C-terminus

Application

Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms
This Anti-Histamine Receptor 1 Antibody, C-terminus is validated for use in ELISA, WB for the detection of Histamine Receptor 1.
Western blot: 1-10 μg/mL using ECL. Recognizes a 57kDa band representing the H1 receptor

ELISA: 1:10,000-1:100,000 using 50-100 ng control peptide per well.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroshi Azuma et al.
The Laryngoscope, 114(12), 2249-2251 (2004-11-27)
Histamine may have physiologic functions in the inner ear. The locations of histamine receptors, however, have not yet been identified in the mammalian cochlea. The aim of this study was to investigate the localization of histamine receptor subtypes (H1, H2
Matthew J Gastinger et al.
The Journal of comparative neurology, 495(6), 658-667 (2006-03-01)
Mammalian retinas are innervated by histaminergic axons that originate from perikarya in the posterior hypothalamus. To identify the targets of these retinopetal axons, we localized histamine receptors (HR) in monkey and rat retinas by light and electron microscopy. In monkeys
Kathryn M Lenz et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 38(37), 8044-8059 (2018-08-11)
Many sex differences in brain and behavior are programmed during development by gonadal hormones, but the cellular mechanisms are incompletely understood. We found that immune-system-derived mast cells are a primary target for the masculinizing hormone estradiol and that mast cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service