Skip to Content
Merck
All Photos(1)

Key Documents

AB5638P

Sigma-Aldrich

Anti-Cannabinoid Receptor 1 Antibody, CT

Chemicon®, from rabbit

Synonym(s):

CB1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€500.00

€500.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€500.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€500.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Cnr1(12801)

General description

CB1 is a protein that is one of two cannabinoid receptors (CB1 CB2). Cannabinoid principally delta-9-tetrahydrocannabinol (THC) and its synthetic analogs, are the active psychoactive compounds present is marijuana. These cannabinoid receptors are members of the guanine-nucleotide-binding protein (G-protein) coupled receptors which inhibit adenylate cyclase activity in a dose-dependent, stereoselective and pertussis toxin-sensitive manner. The two receptors have been found to be involved in the cannabinoid-induced CNS effects (including alterations in mood and cognition) experienced by users of marijuana. Three splice variants have been found for this gene. CB1 is primarily located in brain, and CB2 is found macrophages.

Specificity

Recognizes rat Cannabinoid Receptor 1 (CB1). The immunogen shows no significant sequence homology with CB2 or other receptors.

SPECIES REACTIVITIES: The immunogen peptide is 100% conserved mouse, 94% in monkey and human, 88% in zebra finch, 75% in feline and newt, (salamander) and 73% in puffer fish. Reactivity with other species has not been confirmed.

Immunogen

A 20 amino acid peptide sequence near the cytoplasmic, C-terminus of rat CB1 (Gerard et al. 1991; Gerard et al.

1990; Shire et al. 1995; Matsuda et al. 1990; Ho et al. 1996; Chakraborty et al. 1995).
Epitope: C-terminus

Application

Anti-Cannabinoid Receptor 1 Antibody, C-terminus is an antibody against Cannabinoid Receptor 1 for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Neuroinflammation & Pain

Neurotransmitters & Receptors
Western blot: 1-10 μg/mL using ECL. Expected sizes are ~60kDa and 80-84kDa under reducing conditions.

Immunohistochemistry: not tested. It is recommended that the antibody be tried at 2-10 μg/mL on paraformaldehyde fixed tissue.

ELISA: 1:10,000-1:100,000 using 50-100 ng control peptide (Catalog number AG213) per well.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in phosphate buffered saline containing 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service