Skip to Content
Merck
All Photos(1)

Key Documents

AB5126

Sigma-Aldrich

Anti-Melanocortin Receptor-1 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Mc1R(17199)

Specificity

Melanocyte Stimulating Hormone Receptor (MSH-R), also called Melanotropin Receptor, or Melanocortin-1 Receptor (MC1-R), is a 315 aa transmembrane protein belonging to the family 1 of G-Protein coupled receptors. It is a receptor for MSH (a, b, g) and ACTH. Its activity is mediated by G-proteins that activate adenylate cyclase. It is found in Melanocytes and corticoadrenal tissue as well as various tissues like adrenal gland, leukocytes, lung, lymph node, ovary, testis, pituitary, placenta, spleen and uterus.

Mouse MSH-R. Reactivity with other species has not been confirmed. The immunogen sequence shows 70% homology with human, 66% with bovine, sheep and horse, and 60% with canine.

Immunogen

A 21 mer peptide (designated MCR11) from the 3rd cytoplasmic domain of mouse MC1-R.

Application

Anti-Melanocortin Receptor-1 Antibody is an antibody against Melanocortin Receptor-1 for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Hormones & Receptors
Western blot: 1-10 μg/mL using Chemiluminescence technique

ELISA: 1:10,000 - 100,000



Optimal working dilutions must be determined by end user.

Physical form

Affinity Purified immunoglobulin. Liquid in PBS, 0.1% BSA and 0.05% sodium azide.

Storage and Stability

Can be refrigerated at 2-8°C in undiluted aliquots for short term use. Maintain frozen at -20°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service