Skip to Content
Merck
All Photos(1)

Key Documents

AB15292

Sigma-Aldrich

Anti-Lbx1 Antibody

Chemicon®, from rabbit

Synonym(s):

Ladybird

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€306.00

€306.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€306.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€306.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Lbx1(16814)

Specificity

Lbx1 (Ladybird). By Western blot the antibody recognizes a band at ~30 kDa on mouse spinal cord lysate.

Immunogen

Synthetic peptide.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Neuronal & Glial Markers
This Anti-Lbx1 Antibody is validated for use in WB for the detection of Lbx1.
Western blot: 1:500-1:1,000 on mouse spinal lysate.

Optimal working dilutions must be determined by end user.

Target description

30 kDa

Physical form

Format: Purified
ImmunoAffinity Purified
Liquid in PBS with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service