Skip to Content
Merck
All Photos(1)

Documents

AB1528

Sigma-Aldrich

Anti-Nerve Growth Factor-β Antibody

serum, Chemicon®

Synonym(s):

NGFbeta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

sheep

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human, chicken, mouse

should not react with

bovine

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NGF(4803)

Specificity

Nerve growth factor-beta (NGF). Less than 1% cross-reactivity against recombinant human NT4 and BDNF and approximately 2% for NT3 by ELISA.

Immunogen

Mouse nerve growth factor-beta purified from salivary glands.

Application

Detect Nerve Growth Factor-β using this Anti-Nerve Growth Factor-β Antibody validated for use in ELISA, WB, IH.
Immunohistochemistry at 1:500-1:5,000 (see suggested protocol).

Immunoblotting at 1:500-1:5,000.

ELISA at 1:500-1:5,000.

Inhibition of biological activity in vitro at 1:10-1:50.
BIOLOGICAL ACTIVITY: Neutralizes NGF, but not other neurotrophins.

Use neat for in vivo animal studies (5-10 μL/g body weight).

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Physical form

Whole sheep antisera. Lyophilized. Reconstitute with 250 μL of sterile distilled water.

Storage and Stability

Maintain at -20°C to -70°C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Aridgides et al.
PloS one, 8(2), e57450-e57450 (2013-02-26)
Cardiomyocytes express neurotrophin receptor TrkA that promotes survival following nerve growth factor (NGF) ligation. Whether TrkA also resides in cardiac fibroblasts (CFs) and underlies cardioprotection is unknown. To test whether CFs express TrkA that conveys paracrine signals to neighbor cardiomyocytes
Scott E Counts et al.
Journal of neurochemistry, 113(3), 649-660 (2010-02-06)
Degeneration of locus coeruleus (LC) noradrenergic forebrain projection neurons is an early feature of Alzheimer's disease. The physiological consequences of this phenomenon are unclear, but observations correlating LC neuron loss with increased Alzheimer's disease pathology in LC projection sites suggest
Cheng-Che Lee et al.
Journal of cardiovascular pharmacology, 77(6), 745-755 (2021-06-01)
The NLRP3 inflammasome is activated by myocardial infarction and then induces the activation of inflammatory caspase-1 activation and maturation of IL-1β, a regulator of synthesis of the nerve growth factor (NGF). Here, we studied whether taurine, 2-aminoethanesulphonic acid, can attenuate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service