Skip to Content
Merck
All Photos(1)

Documents

8.52191

Sigma-Aldrich

Fmoc-Lys(Boc)-Thr(psiMe,Mepro)-OH

Novabiochem®

Synonym(s):

Fmoc-Lys(Boc)-Thr(psiMe,Mepro)-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H43N3O8
CAS Number:
Molecular Weight:
609.71
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥97% (TLC)
≥97.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

hydroxyl

storage temp.

2-8°C

InChI

1S/C33H43N3O8/c1-20-27(29(38)39)36(33(5,6)43-20)28(37)26(17-11-12-18-34-30(40)44-32(2,3)4)35-31(41)42-19-25-23-15-9-7-13-21(23)22-14-8-10-16-24(22)25/h7-10,13-16,20,25-27H,11-12,17-19H2,1-6H3,(H,34,40)(H,35,41)(H,38,39)/t20-,26+,27+/m1/s1

InChI key

JAHDJVRHKPYPNY-PHXCCWLDSA-N

General description

This pseudoproline dipeptide is undoubtedly the most effective and simplest to use tool for overcoming aggregation and enhancing peptide quality in Fmoc SPPS for peptide containing the Lys-Thr dipeptide motif. It consists of a dipeptide in which the threonine residue has been reversibly protected as a structure-breaking proline-like TFA-labile oxazolidine. The reason the pseudoproline residue is introduced as a dipeptide is because it avoids the need to acylate the hindered oxazolidine nitrogen. This has the added advantage of extending the peptide chain by two residues in one step. The threonine residue is regenerated during the normal TFA-mediated cleavage reaction.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Overcoming Aggregation in Fmoc SPPS

Linkage

Replaces: 05-20-1116

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in methanol): -19.5 - -15.5 °
Purity (TLC(157A)): ≥ 97 %
Assay (HPLC, area%): ≥ 97.0 % (a/a)
Single impurities (HPLC, area%): ≤ 1.00 % (a/a)
Acetate (IC): ≤ 0.1 %
Ethyl acetate (HS-GC): ≤ 2.0 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

The ease of assembly of a given peptide sequence is hard to predict, which makes peptide synthesis challenging. Review methods and reagents for avoiding aggregation in solid-phase peptide synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service