Skip to Content
Merck
All Photos(1)

Key Documents

8.52114

Sigma-Aldrich

Fmoc-Ile-(Dmb)Gly-OH

Novabiochem®

Synonym(s):

Fmoc-Ile-(Dmb)Gly-OH, N-α-Fmoc-L-Isoleucyl-N-α-(2,4-dimethoxybenzyl)-glycine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€249.00
5 G
€1,030.00

€249.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
1 G
€249.00
5 G
€1,030.00

About This Item

Empirical Formula (Hill Notation):
C32H36N2O7
Molecular Weight:
560.64
UNSPSC Code:
12352209
NACRES:
NA.22

€249.00


In StockDetails


Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥90.0% (acidimetric)
≥96.0% (HPLC)
≥98% (TLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

General description

Fmoc-Aaa-(Dmb)Gly-OH dipeptides offer the same benefits as pseudoproline dipeptides in Fmoc SPPS but for peptide sequences containing Gly. They are extremely easy to use. Standard coupling methods like PyBOP® /DIPEA or DIPCDI/HOBt can be used for their introduction. Removal of the Dmb group and regeneration of the glycine residue occurs during the course of standard TFA-mediated cleavage reaction.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Overcoming Aggregation in Fmoc SPPS

Linkage

Replaces: 04-12-1280

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in methanol): -34.0 - -28.0 °
Purity (TLC(157A)): ≥ 98 %
Assay (HPLC, area%): ≥ 96.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 90.0 %
Water (K. F.): ≤ 1.50 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
PyBOP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

The ease of assembly of a given peptide sequence is hard to predict, which makes peptide synthesis challenging. Review methods and reagents for avoiding aggregation in solid-phase peptide synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service