Skip to Content
Merck
All Photos(1)

Documents

8.52096

Sigma-Aldrich

Fmoc-Lys(Dabcyl)-OH

≥97% (TLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-Lys(Dabcyl)-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H37N5O5
CAS Number:
Molecular Weight:
619.71
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

product name

Fmoc-Lys(Dabcyl)-OH, Novabiochem®

Quality Level

product line

Novabiochem®

Assay

≥95.5% (HPLC)
≥97% (TLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

InChI

1S/C36H37N5O5/c1-41(2)27-20-18-26(19-21-27)40-39-25-16-14-24(15-17-25)34(42)37-22-8-7-13-33(35(43)44)38-36(45)46-23-32-30-11-5-3-9-28(30)29-10-4-6-12-31(29)32/h3-6,9-12,14-21,32-33H,7-8,13,22-23H2,1-2H3,(H,37,42)(H,38,45)(H,43,44)/t33-/m0/s1

InChI key

FPOPWTDBGMLRNG-XIFFEERXSA-N

General description

A modified lysine derivative for the preparation of chromogenically-labeled peptides by Fmoc SPPS. The Dabcyl group quenches the fluorescence of EDANS, Mca, TET, JOE, FAM fluorophores, making it an extremely useful tool for the synthesis of fluorescence-quenched peptide substrates.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Chromogenic Labeling of Synthetic Peptides

Linkage

Replaces: 04-12-1236

Analysis Note

Color (visual): orange to red
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.0 % (a/a)
Purity (TLC(157B)): ≥ 97 %
Assay (HPLC, area%): ≥ 95.5 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service