Skip to Content
Merck
All Photos(1)

Key Documents

8.41818

Sigma-Aldrich

4-Amino-1-butanol

for synthesis

Synonym(s):

4-Amino-1-butanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11NO
CAS Number:
Molecular Weight:
89.14
MDL number:
UNSPSC Code:
12352104
EC Index Number:
236-364-4

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

206 °C/1013 hPa

mp

16-18 °C

transition temp

flash point 107 °C

density

0.962 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H11NO/c5-3-1-2-4-6/h6H,1-5H2/p+1

InChI key

BLFRQYKZFKYQLO-UHFFFAOYSA-O

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.958 - 0.965
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(C)

104 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service