Skip to Content
Merck
All Photos(1)

Key Documents

8.22322

Sigma-Aldrich

Phosphorus trichloride

for synthesis

Synonym(s):

Phosphorus trichloride, Phosphorus(III) chloride, Phosphorus chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€76.20
500 ML
€104.00
1 L
€180.00
300 KG
€12,270.00

€76.20


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€76.20
500 ML
€104.00
1 L
€180.00
300 KG
€12,270.00

About This Item

CAS Number:
Molecular Weight:
137.33
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-749-3
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
76 °C/1013 hPa
vapor pressure:
131 hPa ( 20 °C)

€76.20


Estimated to ship on31 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

131 hPa ( 20 °C)

Assay

≥99% (argentometric)

form

liquid

potency

18 mg/kg LD50, oral (Rat)

dilution

(for synthesis)

pH

1 (20 °C, 5 g/L in H2O)

bp

76 °C/1013 hPa

mp

-94 °C

density

1.57 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

P(Cl)(Cl)Cl

InChI

1S/Cl3P/c1-4(2)3

InChI key

FAIAAWCVCHQXDN-UHFFFAOYSA-N

General description

for synthesis

Application

Phosphorus trichloride (PCl3) can be used as:
  • A chlorinating agent for the alcohols, aldehydes, ketones, and carboxylic acids.[1]
  • A reagent to convert alcohols, diols, and phenols to phosphites.[1]
  • A reducing agent to reduce sulfoxides, amine N-oxides, and hydroxy amides to corresponding sulfides, amines, and amides.[1]

It can also be used as a reagent to synthesize:
  • Phosphaindolizines and thiazolodiazaphospholes by reacting with quaternary ammonium salts.[1]
  • Phosphorus-containing ligand molecules via the introduction of the phosphorus atom.[1]
  • Benzidronate by treating with phenylacetic acid and phosphorous acid in methanesulfonic acid, followed by hydrolysis.[2]

Analysis Note

Assay (argentometric)≥ 99.0 %Density (d 20 °C/ 4 °C)1.572 - 1.574

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorus (III) Chloride
Moder Kenneth P
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Efficient synthesis of benzidronate applying of phosphorus trichloride and phosphorous acid.
Grun A, et al.
Letters in Drug Design & Discovery, 12(1), 78-84 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service