Skip to Content
Merck
All Photos(1)

Documents

8.22073

Sigma-Aldrich

3-Hydroxytyraminium chloride

for synthesis

Synonym(s):

3-Hydroxytyraminium chloride, 4-(2-Aminoethyl)pyrocatechol hydrochloride, 2-(3,4-Dihydroxyphenyl)ethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3,4-(OH)2C6H3CH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
189.64
MDL number:
UNSPSC Code:
12352107
EC Index Number:
200-527-8

form

powder

potency

2859 mg/kg LD50, oral (Rat)

mp

246-249 °C

density

1.4 g/cm3

storage temp.

2-30°C

InChI

1S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11H,3-4,9H2;1H

InChI key

CTENFNNZBMHDDG-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 99.0 %
Melting range (lower value): ≥ 246 °C
Melting range (upper value): ≤ 249 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service