Skip to Content
Merck
All Photos(1)

Key Documents

8.21003

Sigma-Aldrich

N-Phenyl-1-naphthylamine

for synthesis

Synonym(s):

N-Phenyl-1-naphthylamine, N-1-Naphthylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13N
CAS Number:
Molecular Weight:
219.28
MDL number:
UNSPSC Code:
12352116
EC Index Number:
201-983-0

vapor pressure

0.504 hPa ( 150 °C)

Assay

≥98.0% (GC)

form

solid

autoignition temp.

>500 °C

potency

>2000 mg/kg LD50, oral (Rat)

bp

224 °C/16 hPa

mp

56-60 °C

transition temp

flash point >200 °C

solubility

0.003 g/L

density

1.16 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C16H13N/c1-2-9-14(10-3-1)17-16-12-6-8-13-7-4-5-11-15(13)16/h1-12,17H

InChI key

XQVWYOYUZDUNRW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 56 °C
Melting range (upper value): ≤ 60 °C
Identity (IR): passes test

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B - STOT RE 2

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(C)

> 200 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service