Skip to Content
Merck
All Photos(1)

Key Documents

8.18870

Sigma-Aldrich

1-Bromo-2-chloroethane

for synthesis

Synonym(s):

1-Bromo-2-chloroethane, Ethylene chlorobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4BrCl
CAS Number:
Molecular Weight:
143.41
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-456-0

vapor pressure

4 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

potency

64 mg/kg LD50, oral (Rat)

mp

-17 °C

solubility

7 g/L

density

1.73 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C2H4BrCl/c3-1-2-4/h1-2H2

InChI key

IBYHHJPAARCAIE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.730 - 1.734
Identity (IR): passes test

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service