Skip to Content
Merck
All Photos(1)

Key Documents

8.18734

Sigma-Aldrich

4-Chloro-3,5-dimethylphenol

for synthesis

Synonym(s):

4-Chloro-3,5-dimethylphenol, p-Chloro-m-xylenol

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€54.20

€54.20


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
250 G
€54.20

About This Item

Linear Formula:
4-(Cl)-3,5-(CH3)2C6H2OH
CAS Number:
Molecular Weight:
156.61
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-793-8
NACRES:
NA.22

€54.20


Estimated to ship on02 June 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

crystals

potency

3830 mg/kg LD50, oral (Rat)

pH

7 (0.2 g/L in H2O)

bp

246 °C/1013 hPa

mp

114-116 °C

transition temp

flash point 132 °C

density

0.67 g/cm3

storage temp.

2-30°C

SMILES string

Clc1c(cc(cc1C)O)C

InChI

1S/C8H9ClO/c1-5-3-7(10)4-6(2)8(5)9/h3-4,10H,1-2H3

InChI key

OSDLLIBGSJNGJE-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 112 °C
Melting range (upper value): ≤ 117 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service