Skip to Content
Merck
All Photos(1)

Key Documents

8.18356

Sigma-Aldrich

Hydrogen peroxide urea

for synthesis

Synonym(s):

Hydrogen peroxide urea, Carbamide peroxide powder, Hydrogen peroxide urea powder

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€35.90
500 G
€77.40
10 KG
€710.00
25 KG
€1,440.00

€35.90


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 G
€35.90
500 G
€77.40
10 KG
€710.00
25 KG
€1,440.00

About This Item

Empirical Formula (Hill Notation):
CH4N2O · H2O2
CAS Number:
Molecular Weight:
94.07
MDL number:
UNSPSC Code:
12352120
EC Index Number:
204-701-4
NACRES:
NA.22

€35.90


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: oxidant

pH

6.3 (20 °C in H2O)

mp

75-85 °C (decomposition)

solubility

500 g/L

storage temp.

15-25°C

SMILES string

NC(=O)N.OO

InChI

1S/CH4N2O.H2O2/c2-1(3)4;1-2/h(H4,2,3,4);1-2H

InChI key

AQLJVWUFPCUVLO-UHFFFAOYSA-N

Analysis Note

Assay (iodometric): 33.0 - 37.0 %

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 3 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service