Skip to Content
Merck
All Photos(1)

Key Documents

8.18282

Sigma-Aldrich

Di-tert-butyl dicarbonate

for peptide synthesis

Synonym(s):

Di-tert-butyl dicarbonate

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€42.70
100 ML
€88.90
500 ML
€277.00

€42.70


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
€42.70
100 ML
€88.90
500 ML
€277.00

About This Item

CAS Number:
Molecular Weight:
218.25
MDL number:
UNSPSC Code:
12352108
EC Index Number:
246-240-1
NACRES:
NA.22

€42.70


Estimated to ship on30 May 2025


Request a Bulk Order

Product Name

Di-tert-butyl dicarbonate, for synthesis

Quality Level

form

liquid

autoignition temp.

460 °C

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

bp

56-57 °C/0.7 hPa

mp

20-24 °C

transition temp

flash point 37 °C

density

1.02 g/cm3 at 20 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

O(C(C)(C)C)C(=O)OC(=O)OC(C)(C)C

InChI

1S/C10H18O5/c1-9(2,3)14-7(11)13-8(12)15-10(4,5)6/h1-6H3

InChI key

DYHSDKLCOJIUFX-UHFFFAOYSA-N

General description

for synthesis

Application

Di-tert-butyl dicarbonate for synthesis. CAS 24424-99-5, molar mass 218.25 g/mol.

Analysis Note

Assay (GC, area%)≥ 98.0 % (a/a)Melting range (lower value)≥ 20 °CMelting range (upper value)≤ 24 °CIdentity (IR)passes testDue to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service