Skip to Content
Merck
All Photos(1)

Key Documents

8.18215

Sigma-Aldrich

Tetra-n-butylammonium hydroxide

(12.5% solution in methanol) for synthesis

Synonym(s):

Tetrabutylammonium hydroxide

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€69.30
1 L
€175.00

€69.30


Estimated to ship on31 May 2025



Select a Size

Change View
250 ML
€69.30
1 L
€175.00

About This Item

UNSPSC Code:
12352107
NACRES:
NA.22

€69.30


Estimated to ship on31 May 2025


Quality Level

expl. lim.

5.5-36.5 % (v/v) Methanol)

bp

65 °C/1013 hPa

transition temp

flash point 11 °C

density

0.81 g/cm3 at 20 °C

storage temp.

2-30°C

Application

  • Biological Acid Anion Research: Tetra-n-butylammonium hydroxide is used in the hydration structures of lactic acid, facilitating the formation of ionic clathrate hydrate with biological organic acid anions. This application is significant in the study of biological molecules and their interactions with water molecules in various conditions (Muromachi et al., 2014).
  • Pharmaceutical Analysis: It is used in the determination of calcium dobesilate in human plasma, utilizing ion-pairing extraction and high-performance liquid chromatography. This method is crucial for monitoring drug levels in patients, ensuring therapeutic efficacy and safety (Róna et al., 2001).

Analysis Note

Assay (acidimetric): 11.5 - 13.5 %
Density (d 20 °C/ 4 °C): 0.815 - 0.830

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service