Skip to Content
Merck
All Photos(1)

Key Documents

8.18132

Sigma-Aldrich

2,2,6,6-Tetramethylpiperidine

for synthesis

Synonym(s):

2,2,6,6-Tetramethylpiperidine

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€57.10
100 ML
€131.00

€57.10


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
€57.10
100 ML
€131.00

About This Item

Empirical Formula (Hill Notation):
C9H19N
CAS Number:
Molecular Weight:
141.25
MDL number:
UNSPSC Code:
12352005
EC Index Number:
212-199-3
NACRES:
NA.22

€57.10


Estimated to ship on30 May 2025


Request a Bulk Order

vapor pressure

14 hPa ( 39 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

290 °C

potency

500 mg/kg LD50, oral (Rat)

mp

-59 °C

transition temp

flash point 37 °C

solubility

25.9 g/L

density

0.83 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N1C(CCCC1(C)C)(C)C

InChI

1S/C9H19N/c1-8(2)6-5-7-9(3,4)10-8/h10H,5-7H2,1-4H3

InChI key

RKMGAJGJIURJSJ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.830 - 0.834
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

98.6 °F

Flash Point(C)

37 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service