Skip to Content
Merck
All Photos(1)

Key Documents

8.18129

Sigma-Aldrich

Trimethylsilyl trifluoromethanesulfonate

for synthesis

Synonym(s):

Trimethylsilyl trifluoromethanesulfonate, Trimethylsilyl triflate, Trifluoromethanesulfonic acid trimethylsilyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3CSO2OSi(CH3)3
CAS Number:
Molecular Weight:
222.26
MDL number:
UNSPSC Code:
12352103
EC Index Number:
248-565-4

vapor pressure

4.7 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (acidimetric)

autoignition temp.

405 °C (DIN 51794)

bp

35-37 °C/13 hPa

transition temp

flash point 38 °C (DIN 51755 Part 1)

density

1.23 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H9F3O3SSi/c1-12(2,3)10-11(8,9)4(5,6)7/h1-3H3

InChI key

FTVLMFQEYACZNP-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 1.226 - 1.231
Identity (IR): passes test

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service