Skip to Content
Merck
All Photos(1)

Key Documents

8.14859

Sigma-Aldrich

N-Phenylmorpholine

for synthesis

Synonym(s):

N-Phenylmorpholine, Morpholinobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€51.90

€51.90


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
€51.90

About This Item

Empirical Formula (Hill Notation):
C10H13NO
CAS Number:
Molecular Weight:
163.22
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-164-0
NACRES:
NA.22

€51.90


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

solid

autoignition temp.

410 °C

potency

930 mg/kg LD50, oral (Rat)
360 mg/kg LD50, skin (Rabbit)

bp

260 °C/1013 hPa

mp

48-51 °C

transition temp

flash point 124 °C

density

1.053 g/cm3 at 60 °C

storage temp.

2-30°C

SMILES string

N2(CCOCC2)c1ccccc1

InChI

1S/C10H13NO/c1-2-4-10(5-3-1)11-6-8-12-9-7-11/h1-5H,6-9H2

InChI key

FHQRDEDZJIFJAL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 50 °C
Melting range (upper value): ≤ 54 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service