Skip to Content
Merck
All Photos(2)

Documents

8.14136

Sigma-Aldrich

2-(2-Nitroethyl)-1,3-dioxolane

for synthesis

Synonym(s):

2-(2-Nitroethyl)-1,3-dioxolane, 3-Nitropropionaldehyde ethylene glycol acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO4
CAS Number:
Molecular Weight:
147.13
UNSPSC Code:
12352102

Quality Level

Assay

≥98.0% (GC)

bp

88-90 °C/1 hPa

density

1.25 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.245 - 1.247
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service