Skip to Content
Merck
All Photos(1)

Key Documents

8.14134

Sigma-Aldrich

2-Chloro-3,5-dinitropyridine

for synthesis

Synonym(s):

2-Chloro-3,5-dinitropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H2ClN3O4
CAS Number:
Molecular Weight:
203.54
MDL number:
UNSPSC Code:
12352101
EC Index Number:
219-937-3

Assay

≥99.0% (GC)

form

powder

potency

50 mg/kg LD50, oral (Rat)

mp

62-65 °C

storage temp.

2-30°C

InChI

1S/C5H2ClN3O4/c6-5-4(9(12)13)1-3(2-7-5)8(10)11/h1-2H

InChI key

QLHVJBXAQWPEDI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 62 °C
Melting range (upper value): ≤ 65 °C
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service