Skip to Content
Merck
All Photos(1)

Key Documents

8.06187

Sigma-Aldrich

4-Methylpiperidine

for synthesis

Synonym(s):

4-Methylpiperidine, 4-Pipecoline

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€50.80
500 ML
€176.00

€50.80


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€50.80
500 ML
€176.00

About This Item

Empirical Formula (Hill Notation):
C6H13N
CAS Number:
Molecular Weight:
99.17
MDL number:
UNSPSC Code:
12352005
EC Index Number:
210-954-1
NACRES:
NA.22

€50.80


Estimated to ship on13 April 2025


Request a Bulk Order

vapor pressure

16 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

pH

13 (20 °C, 100 g/L in H2O)

bp

105-108 °C/1013 hPa

mp

4-5 °C

transition temp

flash point 13 °C

density

0.84 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N1CCC(CC1)C

InChI

1S/C6H13N/c1-6-2-4-7-5-3-6/h6-7H,2-5H2,1H3

InChI key

UZOFELREXGAFOI-UHFFFAOYSA-N

Application

  • Alkynyl-naphthalimide Fluorophores: This study presents gold coordination chemistry with alkynyl-naphthalimide fluorophores and their applications in cellular imaging. The research demonstrates the use of 4-Methylpiperidine in the synthesis of these fluorophores, highlighting its role in specialty chemical manufacturing (Langdon-Jones et al., 2015).
  • Palladium-catalyzed synthesis of indole derivatives: 4-Methylpiperidine is used in a palladium-catalyzed one-pot three- or four-component coupling process, proving to be an effective catalyst and pharmaceutical intermediate in the synthesis of indole derivatives, which are significant in organic synthesis and pharmaceutical research (Hao et al., 2014).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.838 - 0.843
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service