Skip to Content
Merck
All Photos(1)

Key Documents

8.02661

Sigma-Aldrich

Cyclooctanol

for synthesis

Synonym(s):

Cyclooctanol

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€47.60

€47.60


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
€47.60

About This Item

Empirical Formula (Hill Notation):
C8H16O
CAS Number:
Molecular Weight:
128.21
MDL number:
UNSPSC Code:
12352104
EC Index Number:
211-800-6
NACRES:
NA.22

€47.60


Estimated to ship on20 May 2025


Request a Bulk Order

vapor pressure

0.1 hPa

Quality Level

Assay

≥95.0% (GC)

form

liquid

autoignition temp.

300 °C

potency

735 mg/kg LD50, oral (Rat)

bp

209 °C/1013 hPa

mp

15 °C

transition temp

flash point 86 °C

solubility

5.4 g/L

density

0.97 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC1CCCCCCC1

InChI

1S/C8H16O/c9-8-6-4-2-1-3-5-7-8/h8-9H,1-7H2

InChI key

FHADSMKORVFYOS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.964 - 0.966
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

186.8 °F

Flash Point(C)

86 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service