Skip to Content
Merck
All Photos(2)

Key Documents

8.02304

Sigma-Aldrich

(-)-Quinine

for resolution of racemates for synthesis

Synonym(s):

(-)-Quinine, 6-Methoxy-α-(5-vinyl-2-quinuclidinyl)-4-quinolinemethanol

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€76.20

€76.20


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
€76.20

About This Item

Empirical Formula (Hill Notation):
C20H24N2O2
CAS Number:
Molecular Weight:
324.42
MDL number:
UNSPSC Code:
12352104
EC Index Number:
205-003-2
NACRES:
NA.22

€76.20


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

form

powder

pH

9.0 (20 °C, 0.5 g/L in H2O)

mp

174-178 °C (decomposition)

solubility

0.5 g/L

bulk density

110‑140 kg/m3

storage temp.

2-30°C

SMILES string

N21[C@@H](C[C@@H]([C@H](C2)C=C)CC1)[C@H](O)c3c4c(ncc3)ccc(c4)OC

InChI

1S/C20H24N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3/t13-,14-,19-,20+/m0/s1

InChI key

LOUPRKONTZGTKE-WZBLMQSHSA-N

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Melting range (lower value): ≥ 174 °C
Melting range (upper value): ≤ 178 °C
Spec. rotation [α²0/D (c=1 in H₂SO₄ 0.1 mol/l): -285.0 - -275.0 °
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service