Skip to Content
Merck
All Photos(2)

Key Documents

8.00077

Sigma-Aldrich

4′-Methylacetanilide

for synthesis

Synonym(s):

4′-Methylacetanilide, N-p-Tolylacetamide, Acetic acid p-toluidide, 4-Acetamidotoluene

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€123.00

€123.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
€123.00

About This Item

Linear Formula:
CH3CONHC6H4-4-(CH3)
CAS Number:
Molecular Weight:
149.19
MDL number:
UNSPSC Code:
12352111
EC Index Number:
203-155-4
NACRES:
NA.22

€123.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

solid

bp

306-308 °C/1013 hPa

mp

147-150 °C

transition temp

flash point 168 °C

storage temp.

2-30°C

SMILES string

N(c1ccc(cc1)C)C(=O)C

InChI

1S/C9H11NO/c1-7-3-5-9(6-4-7)10-8(2)11/h3-6H,1-2H3,(H,10,11)

InChI key

YICAMJWHIUMFDI-UHFFFAOYSA-N

Application

  • Kompleks Ni (II)-4′-Methylacetanilide: Sintesis, Karakterisasi, dan Uji Antibakteri: This study discusses the synthesis, characterization, and antibacterial testing of a nickel (II) complex with 4′-Methylacetanilide. The findings highlight the coordination of the C=O group of 4′-Methylacetanilide to nickel ions (Z Afiifah, 2023).
  • Sintesis, Karakterisasi, dan Uji Antibakteri Kompleks Cu (II) dengan 4′-Methylacetanilide: This paper describes the synthesis and characterization of a copper (II) complex with 4′-Methylacetanilide, focusing on its antibacterial activity against gram-negative bacteria (DF Carolina, 2023).

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 147 °C
Melting range (upper value): ≤ 150 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service