Skip to Content
Merck
All Photos(1)

Key Documents

79173BC

Millipore

GDS PPMX PickPen Tip Reload Rack

BioControl Systems, for use with GDS PickPen® PIPETMAX System

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€1,060.00

€1,060.00


In StockDetails


Request more information

Select a Size

Change View
1 EACH
€1,060.00

About This Item

UNSPSC Code:
41121521

€1,060.00


In StockDetails


Request more information

manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS PickPen® PIPETMAX System

storage temp.

room temp

Legal Information

PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service