Skip to Content
Merck
All Photos(1)

Documents

681660-M

Millipore

N-WASP-Inhibitor, 187-1

The N-WASP-Inhibitor, 187-1 controls the biological activity of neural Wiskott Aldrich syndrome protein (N-WASP). This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

N-WASP-Inhibitor, 187-1, cyclo( LK DF DP DF LF DP LQ)₂

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C96H122N18O16
Molecular Weight:
1784.11
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

DMSO: 1 mg/mL
acetic acid: 5%

storage temp.

−20°C

General description

A 14-amino acid cyclic peptide that potently blocks phosphatidylinositol 4,5-bisphosphate (PIP2)-induced actin assembly in Xenopus egg cytoplasmic extract (IC50 ~ 2 µM). Shown to directly bind to neural Wiskott Aldrich syndrome protein (N-WASP) with high-affinity, and prevent the activation of actin-related protein 2/3 (Arp2/3) complex by inhibiting the interaction between N-WASP and Arp2/3.
A 14-amino acid cyclic peptide that potently blocks phosphatidylinositol 4,5-bisphosphate (PIP2)-induced actin assembly in Xenopus egg cytoplasmic extract (IC50 ~2 µM). Shown to directly bind to neural Wiskott Aldrich syndrome protein (N-WASP) with high-affinity, and allosterically stabilize the autoinhibited conformation of N-WASP. It prevents the activation of actin-related protein 2/3 (Arp2/3) complex by blocking the interaction between N-WASP and Arp2/3.

Biochem/physiol Actions

Primary Target
Neural Wiskott Aldrich syndrome protein (N-WASP)
Target IC50: ~ 2 µM blocking phosphatidylinositol 4,5-bisphosphate (PIP2)-induced actin assembly in Xenopus egg cytoplasmic extract

Warning

Toxicity: Standard Handling (A)

Sequence

cyclo(LLys-DPhe-DPro-DPhe-LPhe-DPro-LGln)₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Peterson, J.R., et al. 2001. Proc. Natl. Acad. Sci. USA98, 10624.
Suetsugu, S., et al. 2001. J. Biol. Chem.276, 33175.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service