Skip to Content
Merck
All Photos(1)

Key Documents

681500

Sigma-Aldrich

WHI-P180

A potent inhibitor of lgE-mediated mast cell responses to allergens in vitro and in vivo.

Synonym(s):

WHI-P180, 4-(3ʹ-Hydroxyphenyl)amino-6,7-dimethoxyquinazoline

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€184.00

€184.00


Estimated to ship on18 April 2025



Select a Size

Change View
1 MG
€184.00

About This Item

Empirical Formula (Hill Notation):
C16H15N3O3
CAS Number:
Molecular Weight:
297.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€184.00


Estimated to ship on18 April 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N(c3cc(ccc3)O)c1ncnc2c1cc(c(c2)OC)OC

InChI

1S/C16H15N3O3/c1-21-14-7-12-13(8-15(14)22-2)17-9-18-16(12)19-10-4-3-5-11(20)6-10/h3-9,20H,1-2H3,(H,17,18,19)

InChI key

BNDYIYYKEIXHNK-UHFFFAOYSA-N

General description

A potent inhibitor of IgE-mediated mast cell responses to allergens in vitro and in vivo. Also inhibits cyclin-dependent kinase 2 (Cdk2; IC50 = 1 µM) by blocking the ATP site.
A potent inhibitor of lgE-mediated mast cell responses to allergens in vitro and in vivo. Also inhibits cyclin-dependent kinase 2 (Cdk2; IC50 = 1 µM) by blocking the ATP site.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Cdk2
Product does not compete with ATP.
Reversible: no
Target IC50: 1 µM for Cdk2

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Shewchuk, L., et al. 2000. J. Med. Chem. 43, 133.
Chen, C.L., et al. 1999. Pharm. Res. 16, 117.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service