Skip to Content
Merck
All Photos(1)

Key Documents

660

Sigma-Aldrich

May-Grünwald Stain

Harleco®, suitable for hematology

Synonym(s):

Hematology Stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
EC Index Number:
200-659-6

form

liquid

Quality Level

manufacturer/tradename

Harleco®

mp

-98 °C

density

0.791 g/cm3

λmax

520-530 nm (Eosin)
647-657 nm (Azure)

application(s)

diagnostic assay manufacturing
hematology
histology

hematology ((contains Methanol))

shipped in

ambient

storage temp.

15-25°C

General description

HARLECO® Blood stain producing results similar to Wright Stain. May be used in conjunction with Giemsa Stain for bone marrow preparations

Application

HARLECO® Blood stain producing results similar to Wright Stain. May be used in conjunction with Giemsa Stain for bone marrow preparations
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Blood stain producing results similar to Wright Stain. May be used in conjunction with Giemsa Stain for bone marrow preparations.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service