Skip to Content
Merck
All Photos(1)

Key Documents

6310-OP

Millipore

MOPS

>=99.0%, crystalline powder (Fine), OmniPur® Grade

Synonym(s):

OmniPur® MOPS, 3-(N-Morpholino)propanesulfonic acid, 4-Morpholinepropanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

Product Name

MOPS, OmniPur® Grade, >=99.0%

Quality Level

form

crystalline powder (Fine)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

InChI

1S/C7H15NO4S/c9-13(10,11)7-1-2-8-3-5-12-6-4-8/h1-7H2,(H,9,10,11)

InChI key

DVLFYONBTKHTER-UHFFFAOYSA-N

General description

Molecular biology grade.

Specifications:
A (260 nm, 0.1 M, water): ≤0.05AU
A (280 nm, 0.1 M, water): ≤0.05AU
Assay: ≥99.0%
DNase: None detected
Phosphatase: None detected
pKa at 25°C: 6.50-7.90
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service