Skip to Content
Merck
All Photos(1)

Documents

616395

Sigma-Aldrich

Transferrin, Apo-, Human Plasma

Synonym(s):

Transferrin, Apo-, Human Plasma

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.25

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

pI 

5.8

capacity

1.20-1.60 μg/mg binding capacity (iron)(total)

solubility

water: 10 mg/mL

cation traces

Fe: <0.01%

shipped in

ambient

storage temp.

−20°C

General description

Native apo-transferrin from human plasma. Transferrin is a major iron binding serum protein found in human plasma or serum at concentrations of 3 mg/ml. Its concentration correlates with the total iron binding capacity of serum and its level is regulated by the availability of iron (total iron binding capacity: 1.20-1.60 μg/mg). It has bacteriostatic and fungistatic activity and is essential for iron utilization by the hemin-synthesizing reticulocytes. Stored in bone marrow as Tf-bound iron. Has a pI of 5.8.
Native apo-transferrin from human plasma. Transferrin is a serum protein involved in iron binding and transport. Substantially iron-free.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 10 mM sodium phosphate buffer, pH 7.4.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 2 weeks at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service