Skip to Content
Merck
All Photos(1)

Key Documents

6.04760

Supelco

Iodine

resublimed

Synonym(s):

Iodine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I2
CAS Number:
Molecular Weight:
253.81
MDL number:
UNSPSC Code:
12141916
Pricing and availability is not currently available.

SMILES string

II

InChI

1S/I2/c1-2

InChI key

PNDPGZBMCMUPRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • DL-Serine Hydrazide Hydrochloride Multiple-site Synergy Induced Effective and Stable Formamidine-Rich Perovskite Solar Cells.: This research explores the use of iodine in enhancing the stability and efficiency of perovskite solar cells, a promising development for renewable energy technologies (Gong et al., 2024).

  • Dynamic Cleavage-Remodeling of Covalent Organic Networks into Multidimensional Superstructures.: Investigates the role of iodine in the dynamic remodeling of covalent organic frameworks, offering insights into the creation of advanced materials with tunable properties (Jiang et al., 2024).

  • Machine Learning for Characterizing Biofuels Based on Femtosecond Laser Ionization Mass Spectrometry.: Details the application of iodine as a dopant in mass spectrometry for enhancing the detection and characterization of biofuels, illustrating the integration of advanced analytical techniques and machine learning (Imasaka et al., 2024).

  • Indoor concentrations of radioactive aerosols from nuclear accidents.: Discusses the critical role of iodine in monitoring and mitigating the effects of radioactive aerosols in nuclear accident scenarios, emphasizing its importance in environmental safety and public health (Orabi, 2024).

  • Preoperative prediction of lymphovascular invasion and T-staging of rectal cancer via a dual-energy computed tomography iodine map: a feasibility study.: Highlights the use of iodine-based imaging for preoperative assessments in rectal cancer, providing a non-invasive method to improve diagnostic accuracy and patient management (Zhang et al., 2024).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Respiratory system, Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service